α-Cyclodextrin ≥98.0% (by HPLC)
Glass bottle, 25 g
Catalog # TCIAC0776-25G
Supplier: TCI
Jump to:
Product Details & Documents
- CAS Number:10016-20-3
- Formula:C₃₆H₆₀O₃₀
- MDL Number:MFCD00078207
- Molecular Weight:972.84 g/mol
- Merck Index:14,02718
- EINECS:233-007-4
Specifications
Specification Test Results
- Appearance:White powder to crystal
- Purity (HPLC):min. 98.0 area%
- Specific rotation [a]20/D:+146.0 to +153.0 deg (C=1, H2O) (calcd.on anh.substance)
- Solubility in Water:almost transparency
- Water:max. 10.0 %
Specifications
- Pack type:Glass bottle
- Pk:25 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



