Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Phenol red

Glass bottle, 1 g
  TCIAP0100-1G
 
 :   TCI
Phenol red
  TCIAP0100-1G
 :  TCI
 :  143-74-8
Phenol red
Phenol red

Glass bottle, 1 g

 :  143-74-8
 
17,60 €
   
1 * 1 g
 
 :  3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
  •  :143-74-8
  •  :C₁₉H₁₄O₅S
  •  :MFCD00003552
  •  :354.38 g/mol
  •  :14,07247
  •  :> 300 °C
  •  :Ambient
  •  :205-609-7
 

 

  • Appearance:
    Dark red to Brown powder to crystaline
  • Absorbance:
    min. 0.530 (5mg/L, pH 8.4, 556.0 to 560.0 nm)
  • Transition interval (pH):
    (yellow)6.8 to 8.4 (red)

 

  • Pack type:
    Glass bottle
  • Pk:
    1 g

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.