Nile red
1 g
Catalog # TCIAN0659-1G
Supplier: TCI
Jump to:
Product Details & Documents
Synonyms: 9-(Diethylamino)-5H-benzo[a]phenoxazin-5-one
- CAS Number:7385-67-3
- Formula:C₂₀H₁₈N₂O₂
- MDL Number:MFCD00011639
- Molecular Weight:318.38 g/mol
- EINECS:230-966-0
Documents
Specifications
Specification Test Results
- Appearance:Yellow to Brown to Dark green powder to crystal
- Lambda max.:550.0 to 554.0 nm (MeOH)
- Absorbance (E1%1cm):min. 1,300 (MeOH, 550.0 to 554.0 nm)
Specifications
- Pk:1 g



