Order Entry
Belgium
Orders LinkContactUsLinkComponent
Copper PAN

Copper PAN

10 g
Catalog # TCIAC0385-10G
Irritant
Supplier:   TCI
Copper PAN
Catalog # TCIAC0385-10G
Supplier:  TCI
CAS Number:  23299-85-6
Copper PAN

10 g

CAS Number:  23299-85-6
Price
160,00 €
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  Bis[1-(2-pyridylazo)-2-naphtholato]copper(II)
  • CAS Number:23299-85-6
  • MDL Number:MFCD00067176
Specifications

Specification Test Results

  • Appearance:
    Orange to Amber to Dark red powder to crystal
  • Absorbance:
    0.500 to 0.550 (50 %, Dioxane, 466.0 to 472.0 nm)

Specifications

  • Pk:
    10 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.