Vials
Vials, LP, with write on label, 2,00 ml, Clear, No
Catalog # HEWL5190-4017
Supplier: Agilent
Jump to:
Product Details & Documents
Specifications
- Cap included:No
- Capacity:2,00 ml
- Colour:Clear
- Description:Vials, LP, with write on label
- Seq:29