Peristaltic pumps, FlexiPump®
Product Details & Documents
FlexiPump® is a range of peristaltic pumps for culture media preparation. Use it for serial dispensing of culture media, agar or any liquid diluent for efficient sample preparation.
- Select from 3 dispensing modes: Continuous, dose (predetermined volume), multi-dose (predetermined volume, with an adjustable pause between each dose)
- 32 editable programs
- Rotating button for easy navigation
- Compact: Only 16 cm wide
- Precise flow control: Up to 1% precision
- Dispenses liquid from 50 μl to 99 L
- Contamination free: Dispensing assembly is entirely autoclavable with bottle
- Anti-drop system
- Easy dispensing
- Improved dispensing control with the handy gun and the adjustable arm
- Maintenance-free
- High quality Watson Marlow™ pump heads
Available in 2 models:
FlexiPump® with the essential functions
FlexiPump® Pro for high-performance dispensing
FlexiPump® is delivered with 1 peristaltic Watson Marlow™ pump head(s), 1 set of 3,2 mm dispensing assembly (nozzle + GL 45 bottle connector + filter + tubing +
tube weight), user's manual, power cord, free sterile packs of filter bags.
FlexiPump® Pro is delivered with 2 peristaltic Watson Marlow™ pump head(s), 1 set of 3,2 mm dispensing assembly (nozzle + GL 45 bottle connector + filter +
tubing + tube weight), 1 set of 6,4 mm dispensing assembly, user's manual, power cord, USB cable, free sterile packs of filter bags.
: In compliance with ISO 6887-1 and ISO 7218 standards
Specifications
- Désignation:FlexiPump® Pro peristaltic pump, with double pump head and 6 rollers, for culture preparation
- Seq:11
- Débit min.:3 ml/min
- Cat. No.:INSC562000
- Ø intérieur tuyau:1,6 − 3,2 − 4,8 −6,4 − 8 mm
- Poids:5,4 kg
- Matériau du boîtier:Stainless steel
- Débit max.:2500 ml/min
- L×P×H:155×320×210 mm
- Vitesse de rotation:10−550 rpm
- Exigence électrique:100−240 V / 50−60 Hz
- Modèle:FlexiPump® Pro
Accessories
Product Information
- Catalog #DésignationAvailabilityPrice
- INSC561002Dispensing arm, adjustable, for use with other containersLog in for availability1 * 1 ST - 374,00 €
Product Information
- Catalog #DésignationAvailabilityPrice
- 710-1887Handy gun, dispensing control with handy gun including electronic start buttonLog in for availability1 * 1 ST - 362,00 €
Product Information
- Catalog #DésignationAvailabilityPrice
Product Information
- Catalog #DésignationAvailabilityPrice
Product Information
- Catalog #DésignationAvailabilityPrice
Product Information
- Catalog #DésignationAvailabilityPrice
Product Information
- Catalog #DésignationAvailabilityPrice
- INSC5130115 sets of silicone tubes for pump part (200 mm each), inner Ø 4,8 mmLog in for availability1 * 1 SET - 78,00 €



