Peristaltic pumps, FlexiPump®
FlexiPump® is a range of peristaltic pumps for culture media preparation. Use it for serial dispensing of culture media, agar or any liquid diluent for efficient sample preparation.
- Select from 3 dispensing modes: Continuous, dose (predetermined volume), multi-dose (predetermined volume, with an adjustable pause between each dose)
- 32 editable programs
- Rotating button for easy navigation
- Compact: Only 16 cm wide
- Precise flow control: Up to 1% precision
- Dispenses liquid from 50 μl to 99 L
- Contamination free: Dispensing assembly is entirely autoclavable with bottle
- Anti-drop system
- Easy dispensing
- Improved dispensing control with the handy gun and the adjustable arm
- Maintenance-free
- High quality Watson Marlow™ pump heads
Available in 2 models:
FlexiPump® with the essential functions
FlexiPump® Pro for high-performance dispensing
FlexiPump® is delivered with 1 peristaltic Watson Marlow™ pump head(s), 1 set of 3,2 mm dispensing assembly (nozzle + GL 45 bottle connector + filter + tubing +
tube weight), user's manual, power cord, free sterile packs of filter bags.
FlexiPump® Pro is delivered with 2 peristaltic Watson Marlow™ pump head(s), 1 set of 3,2 mm dispensing assembly (nozzle + GL 45 bottle connector + filter +
tubing + tube weight), 1 set of 6,4 mm dispensing assembly, user's manual, power cord, USB cable, free sterile packs of filter bags.
Certifications: In compliance with ISO 6887-1 and ISO 7218 standards
Documents
- Description:FlexiPump® peristaltic pump, with single pump head and 3 rollers, for culture preparation
- Seq:01
- Min. flow rate:1,5 ml/min
- Cat. No.:INSC561000
- Int. Ø tubing:1,6 − 3,2 − 4,8 −6,4 − 8 mm
- Rotation speed:10−550 rpm
- Model:FlexiPump®
- W×D×H:155×270×210 mm
- Max. flow rate:1250 ml/min
- Electrical:100−240 V / 50−60 Hz
- Housing material:Stainless steel
- Weight:5,2 kg
Accessories
- Description
- Description
- 710-1887Handy gun, dispensing control with handy gun including electronic start button1 * 1 ST - 362,00 €
- Description
- Description
- Description
- Description
- Description
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



