ContactUsLinkComponent
15 COMPOSES 100 uG/ML CHAQUE B Ge Hf Mo Nb P Re S Sb Si Sn Ta Ti W Zr DANS HNO3 2% HF 0.2% 1 * 125 mL
Catalog # 89214.200
15 COMPOSES 100 uG/ML CHAQUE B Ge Hf Mo Nb P Re S Sb Si Sn Ta Ti W Zr DANS HNO3 2% HF 0.2% 1 * 125 mL
Catalog # 89214.200
Frequently Bought Together