Order Entry
Belgium
Orders LinkContactUsLinkComponent
Heating tapes

Heating tapes

Heating tape, 2,0 m length, 500 W
Catalog # WINKWBG00402000500
Supplier:   WINKLER
Heating tapes
Catalog # WINKWBG00402000500
Supplier:  WINKLER
Heating tapes
Heating tapes

Heating tape, 2,0 m length, 500 W

Price
Inquire For Price
Log in for availability
Product Details & Documents
Specifications
  • Description:
    Heating tape, 2,0 m length, 500 W
  • Seq:
    04
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.