Order Entry
Belgium
Orders LinkContactUsLinkComponent
Heating tapes

Heating tapes

Heating tape, 1,8 m length, 200 W
Catalog # WINKWBS00201800200
Supplier:   WINKLER
Heating tapes
Catalog # WINKWBS00201800200
Supplier:  WINKLER
Heating tapes
Heating tapes

Heating tape, 1,8 m length, 200 W

Price
Inquire For Price
Log in for availability
1 * 1 Pce
Product Details & Documents

These heating tapes contain four heat conductors, insulated silicone, outer braiding with tinned copper wire, factory terminated and vulcanised main connection.

  • Very high and constant heating power per meter
  • High operating temperature
  • Extremely flexible
  • Quick and easy installation
Specifications
  • Description:
    Heating tape, 1,8 m length, 200 W
  • Seq:
    06
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.