Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Heating tapes
Heating tape, 7,3 m length, 550 W
  WINKWBS00207300550
 :   WINKLER
Heating tapes
  WINKWBS00207300550
 :  WINKLER
Heating tapes
Heating tapes
Heating tape, 7,3 m length, 550 W
1 * 1 Pce
These heating tapes contain four heat conductors, insulated silicone, outer braiding with tinned copper wire, factory terminated and vulcanised main connection.
  • Very high and constant heating power per meter
  • High operating temperature
  • Extremely flexible
  • Quick and easy installation
  • Désignation:
    Heating tape, 7,3 m length, 550 W
  • Seq:
    16