ContactUsLinkComponent
Buffer pack PCR Phusion GC detergent-free final 1x buffer concentration equals 1,5mM MgCl2, contains 4x1,5ml of 5x buffer, 2x1,5ml o f 50mM MgCl2 solution, 1x0,5ml of 100% DMSO 1 * 1 KIT
Catalog # F-521L
Supplier:  FINNZYMES REAGENTS
Buffer pack PCR Phusion GC detergent-free final 1x buffer concentration equals 1,5mM MgCl2, contains 4x1,5ml of 5x buffer, 2x1,5ml o f 50mM MgCl2 solution, 1x0,5ml of 100% DMSO 1 * 1 KIT
Catalog # F-521L
Supplier:  FINNZYMES REAGENTS
Frequently Bought Together