ContactUsLinkComponent
DNA Polymerase DyNAzyme EXT (1U/µl) supplied with 10x DyNAzyme EXT Optimized Buffer, 50mM MgCl2-solution and DMSO. Amplifies fragmen ts up to 40 kb. 1 * 1 KIT
Catalog # F-505L
Supplier:  FINNZYMES REAGENTS
DNA Polymerase DyNAzyme EXT (1U/µl) supplied with 10x DyNAzyme EXT Optimized Buffer, 50mM MgCl2-solution and DMSO. Amplifies fragmen ts up to 40 kb. 1 * 1 KIT
Catalog # F-505L
Supplier:  FINNZYMES REAGENTS
Frequently Bought Together