Order Entry
Belgium
Orders LinkContactUsLinkComponent
Rectangular cover glasses

Rectangular cover glasses

Coverslips, 9×35 mm, 0,13 - 0,16 mm
N° de catalogue BELC1916-09035A
Marketsource
Fournisseur:   BELLCO GLASS
Rectangular cover glasses
N° de catalogue BELC1916-09035A
Fournisseur:  BELLCO GLASS
Marketsource
Rectangular cover glasses
Rectangular cover glasses

Coverslips, 9×35 mm, 0,13 - 0,16 mm

Prix
53,67 €
Log in for availability
1 * 100 ST
Product Details & Documents

These rectangular cover glasses are designed for use with culture tubes.

Specifications
  • Description:
    Coverslips
  • L×W:
    9×35 mm
  • Seq:
    001
  • Thickness:
    0,13 - 0,16 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.