Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Ball valves with thread

15 mm, 12,7 mm inner inflowing/M29×1,75 outer outflowing
  BURK0500-8102
 :   BURKLE
Ball valves with thread
  BURK0500-8102
 :  BURKLE
Ball valves with thread
Ball valves with thread

15 mm, 12,7 mm inner inflowing/M29×1,75 outer outflowing

 
30,00 €
   
1 * 1 ST
 

Ball valve made of PP with outer/inner thread.

  • Maximum operating pressure 10 bar (at +20 °C)
  • Maximum operating temperature 100 °C
  • Suitable for food

It also consists fibre-glass reinforced, ball made of PP, handle with O-ring made of food-safe NBR and a seal made of HDPE.

 
  • Hole Ø:
    15 mm
  • Thread:
    12,7 mm inner inflowing/M29×1,75 outer outflowing
  • Seq:
    01
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.