Orders LinkContactUsLinkComponent
Cryoboxes
Cryoboxes
4×4, 50 ml tubes, —, - mm
Catalog # HEATHS2860P
Supplier:   HEATHROW
Cryoboxes
Catalog # HEATHS2860P
Supplier:  HEATHROW
Cryoboxes
Cryoboxes
4×4, 50 ml tubes, —, - mm
Price
58,70 €
Log in for availability
1 * 10 ST
Product Details & Documents
Made of cardboard, white.
  • Write-on surface for descriptions or self numbering systems
  • Dividers are flat packed and comes in two different sizes (to be ordered separately)
For storage of large samples and 15 or 50 ml tubes.
Documents
Specifications
  • Compartments:
    4×4
  • For:
    50 ml tubes
  • W×D×H ext.:
    - mm
  • Colour:
  • Seq:
    02