Test panels
Test panel, aluminium, matt, chromated alodine, 75×150×0,8 mm
Catalog # SIMEVF8524
Supplier: Industrial Physics
Jump to:
Product Details & Documents
Test panels are used to enhance reproducibility of physical and chemical tests. Each panel is equipped with a hole for hanging and handling.
- Consistent and reproducible quality with sophisticated finishing: No burrs or sharp edges
- Physical testing: Adhesion, elongation, impact resistance and elasticity
- Appearance testing of coatings (gloss, colour, texture)
Documents
TQC Company Video
Vidéos
Specifications
- Description:Test panel, aluminium, matt, chromated alodine
- Seq:11
- W×D×H:75×150×0,8 mm
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.