Order Entry
Belgium
Orders LinkContactUsLinkComponent
Temperature probes

Temperature probes

Liquid probe, Type 80104, 0…1100 °C, 3×130 mm
Catalog # SIMETE5104
Supplier:   Industrial Physics
Temperature probes
Catalog # SIMETE5104
Supplier:  Industrial Physics
Temperature probes
Temperature probes

Liquid probe, Type 80104, 0…1100 °C, 3×130 mm

Price
187,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

These temperature probes can be connected to instruments suited for changeable probes (type ‘K’) using a plug connection.

  • Provided with a firm hand-grip and a one meter curled cable
  • Thicker probes have a longer response time

Documents

Specifications
  • Range:
    0…1100 °C
  • Description:
    Liquid probe
  • Seq:
    02
  • Type:
    Type 80104
  • Ø×L:
    3×130 mm