Inspection sets
Product Details & Documents
These basic sets are designed to perform all measurements and tests regarding painting.
Delivery information: Moisture meter, digital thermo/hygrometer, compass, illuminated surface microscope (to check the surface and paint layer), stainless steel cutter knife, pH-indicator paper (to check the acidity of concrete walls and floors), wedge (for checking reverse-side tolerance and thickness of glass), telescopic inspection mirror and lamp (for visual control), waterproof marker and MasterPaintPlate (multifunctional instrument to measure wet film thickness, cover of edges, thickness of glass, as well as providing an indication of the viscosity and adhesion of the coating).
Documents
TQC Company Video
Videos
Specifications
- Description:Inspection set with non-destructive wood moisture meter
- Seq:002
Accessories
Product Information
- Catalog #AvailabilityPrice
- Knife with rubber grip
Product Information
- Catalog #AvailabilityPrice
- Sturdy stainless steel bradle for dry rot inspection in timberSturdy stainless steel bradle for dry rot inspection in timberSIMESP6202Log in for availability1 * 1 ST - 74,40 €
Product Information
- Catalog #AvailabilityPrice
- Water resistant marker