Test charts
Test charts, White/black, A6, 105×148 mm
Catalog # SIMEVF2317
Supplier: Industrial Physics
Jump to:
Product Details & Documents
Test charts are used for testing physical properties of coating, lacquers, and inks.
- Dimensions from A6 up to A4
- Film laminated for an excellent solvent, chemical resistance, and film spread
- Section for the date, time, and test number
Suitable for determining hiding power, opacity, and spreading rate in coating laboratories and paint production.
: A certificate is available for charts with an optical brightener.
Documents
Specifications
- Format:A6
- Colour:White/black
- Description:Test charts
- Seq:01
- W×H:105×148 mm