Orders LinkContactUsLinkComponent
Surface roughness comparators
Surface roughness comparators
Steel surface roughness comparator for shot blasting, Profiles: 25, 40, 70, 100
Catalog # SIMELD2050
Supplier:   Industrial Physics
Surface roughness comparators
Catalog # SIMELD2050
Supplier:  Industrial Physics
Surface roughness comparators
Surface roughness comparators
Steel surface roughness comparator for shot blasting, Profiles: 25, 40, 70, 100
Price
204,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Quality steel comparison standards designed to indicate the surface condition of blasted steel, in grades of fine, medium and coarse.
  • High purity electroformed nickel reference plates
  • Copied from a certified mild steel master
  • Protected test surfaces when not in use
Certifications: ASTM D4417, ISO 8503
Delivery information: Supplied in a sturdy leather pouch.
Documents
Specifications
  • Model:
    Profiles: 25, 40, 70, 100
  • Description:
    Steel surface roughness comparator for shot blasting