Orders LinkContactUsLinkComponent
Surface profile comparators
Surface profile comparators
Surface profile comparator for grit
Catalog # SIMELD2052
Supplier:   Industrial Physics
Surface profile comparators
Catalog # SIMELD2052
Supplier:  Industrial Physics
Surface profile comparators
Surface profile comparators
Surface profile comparator for grit
Price
297,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Surface comparators for determining roughness by touch and sight.
  • Five different grades of roughness in each comparator
  • Designed to be used with the illuminated magnifier
  • Available in Imperial units only
Section profiles: 0,5 ; 1,0 ; 2,0 ; 3,0 ; 4,0 mils for the three versions.
Certifications: ASTM D4417
Documents
Specifications
  • Description:
    Surface profile comparator for grit
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Surface profile disc holder and magnifier (5×)
    Surface profile disc holder and magnifier (5×)
    SIMELD2054
    Log in for availability
    1 * 1 ST - 125,00 €