Orders LinkContactUsLinkComponent
Surface roughness comparators
Surface roughness comparators
Surface roughness comparator for grit blasting, Profiles: 25, 60, 100, 150
Catalog # SIMELD2040
Supplier:   Industrial Physics
Surface roughness comparators
Catalog # SIMELD2040
Supplier:  Industrial Physics
Surface roughness comparators
Surface roughness comparators
Surface roughness comparator for grit blasting, Profiles: 25, 60, 100, 150
Price
220,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Quality steel comparison standards designed to indicate the surface condition of blasted steel, in grades of fine, medium and coarse.
  • High purity electroformed nickel reference plates
  • Copied from a certified mild steel master
  • Protected test surfaces when not in use
Certifications: ASTM D4417, ISO 8503
Delivery information: Supplied in a sturdy leather pouch.
Documents
Specifications
  • Model:
    Profiles: 25, 60, 100, 150
  • Description:
    Surface roughness comparator for grit blasting