Orders LinkContactUsLinkComponent
Pretreatment test sets
Pretreatment test sets
Pretreatment test set, Basic
Catalog # SIMESP7315
Supplier:   Industrial Physics
Pretreatment test sets
Catalog # SIMESP7315
Supplier:  Industrial Physics
Pretreatment test sets
Pretreatment test sets
Pretreatment test set, Basic
Price
783,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents
The quality of pretreatment prior to application of the coating is essential to the quality of the coating. This pretreatment set is designed for all the parameters during the pretreatment of steel prior to painting.
Certifications: ISO 8502-6 and 9, IMO MSC.215 (82), IMO MSC.244 (83), ISO 8502-3, ISO 8502-4, BS 7079-B4, US Navy NSI 009-32, US Navy PPI 63101-000, ASTM D4417 – B, ISO 2808-3, ASTM D4138, ISO 8501-1:2007, ISO 8503 1 and 2, ASTM D 4417 Method A, ASTM E2501, ISO 2409 :2003 and ASTM D3359.
Delivery information: Basic set contains a Bresle test for surface and media measurements, a dust set and telescopic inspection mirror.
Full set contains Dewcheck four dew point gauge, coating adhesion tester master paint plate, surface profile and coating thickness gauge, spring loaded roller, blast cleanliness standard books, grit profile comaprator and UV inspection flashlight.
Documents
Specifications
  • Model:
    Basic
  • Description:
    Pretreatment test set
  • Seq:
    001