Orders LinkContactUsLinkComponent
Wire baskets, square
Wire baskets, square
118×118×144 mm
Catalog # DELTH-200
Supplier:   DELTALAB
Wire baskets, square
Catalog # DELTH-200
Supplier:  DELTALAB
Wire baskets, square
Wire baskets, square
118×118×144 mm
Price
161,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Square baskets, made of stainless steel wire, with folding handles.
  • Features a strong woven mesh base
  • Good chemical resistance
  • Good temperature resistance
  • Comes without lid (to order baskets with lid please contact our customer service
Documents
Specifications
  • Seq:
    01
  • W×D×H:
    118×118×144 mm