Order Entry
Belgium
Orders LinkContactUsLinkComponent
Tanks, rectangular, LDPE, LaboPlast®

Tanks, rectangular, LDPE, LaboPlast®

85 L, Seamless containers, rectangular shaped, 600×340×480 mm
Catalog # BURK3440-0855
Supplier:   BURKLE
Tanks, rectangular, LDPE, LaboPlast®
Catalog # BURK3440-0855
Supplier:  BURKLE
Tanks, rectangular, LDPE, LaboPlast®
Tanks, rectangular, LDPE, LaboPlast®

85 L, Seamless containers, rectangular shaped, 600×340×480 mm

Price
432,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

These seamless rectangular tanks feature optimum ratio of wall thickness to filling load.

  • Linear LDPE, transparent
  • Rupture-proof
Specifications
  • Capacity:
    85 L
  • Description:
    Seamless containers, rectangular shaped
  • Seq:
    06
  • W×D×H:
    600×340×480 mm
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Container thread ¾" for cylindrical containers
    Container thread ¾" for cylindrical containers
    BURK3440-0034
    Log in for availability
    1 * 1 ST - 213,00 €