Orders LinkContactUsLinkComponent
Drawer system, SOLOBOX
Drawer system, SOLOBOX
320×410×220 mm
Catalog # 216-1054
Supplier:   MULTIROIR
Drawer system, SOLOBOX
Catalog # 216-1054
Supplier:  MULTIROIR
Drawer system, SOLOBOX
Drawer system, SOLOBOX
320×410×220 mm
Price
67,50 €
Log in for availability
1 * 1 ST
Product Details & Documents
White PP frame, translucent drawers.
  • Stackable
  • Impact resistant
SOLOBOX is a modular individual drawer system. For well organised storage of many items, on shelves or at the workstation.
Specifications
  • Seq:
    05
  • W×D×H:
    320×410×220 mm