Order Entry
Belgium
Orders LinkContactUsLinkComponent
LIGHTBOX Multipurpose Boxes, with Lid

LIGHTBOX Multipurpose Boxes, with Lid

5 L, 334×190×110 mm, PP
Catalog # 216-1048
Supplier:   MULTIROIR
LIGHTBOX Multipurpose Boxes, with Lid
Catalog # 216-1048
Supplier:  MULTIROIR
LIGHTBOX Multipurpose Boxes, with Lid
LIGHTBOX Multipurpose Boxes, with Lid

5 L, 334×190×110 mm, PP

Price
11,20 €
Log in for availability
1 * 1 ST
Product Details & Documents

Discover this range of polypropylene storage boxes with lids, suitable for storing and organising consumables and laboratory equipment.

  • Available in different sizes
  • Resistant to common chemicals
  • Durable against shocks and frequent handling
  • Stable under temperature variations
  • Secure lids for optimal protection of samples
  • Versatile use for various types of laboratory equipment
  • Easy to clean and decontaminate

Optimise laboratory organisation with this range of polypropylene storage boxes, specifically designed to meet the demands of scientific environments. These sturdy boxes are equipped with secure lids and provide an ideal solution for storing and organising sensitive laboratory consumables and equipment.

Made from high-quality polypropylene, these boxes are resistant to common chemicals, temperature variations, and shocks, ensuring optimal protection for samples and reagents. Their transparency allows for quick identification of contents, while the stackable design maximises space utilisation on benches and in storage cabinets.

Specifications
  • Capacity:
    5 L
  • Material:
    PP
  • Seq:
    01
  • W×D×H:
    334×190×110 mm