Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
CONTROL STANDARD ENDOTOXIN E.COLI O113:H10, USE WITH PYROCHROME KITS, 10ng/VIAL, 5 VIALS 1 * 5 items
Catalog # CAPEEC010-5
Supplier:
Associates of Cape Cod
Expand image
CONTROL STANDARD ENDOTOXIN E.COLI O113:H10, USE WITH PYROCHROME KITS, 10ng/VIAL, 5 VIALS 1 * 5 items
Catalog # CAPEEC010-5
Supplier:
Associates of Cape Cod
Related Information
Documentation
CoC_CAPEEC010-5_249034
Documentation
Frequently Bought Together
Antimicrobial susceptibility disks
Supplier: THERMO FISHER DIAGNOSTICS
TEST IDENTIFICAZ./CLASSIFICA- ZIONE 1 * 1 items
Supplier: LIOFILCHEM
Culti-Loops™ <i>Haemophilus influenzae</i> ATCC™ 35540™
Supplier: THERMO FISHER DIAGNOSTICS
Phyto Pspi Agar Base (SNAC Medium) 1 * 500 g
Supplier: HIMEDIA
VWR® Culture Media, Ready to Use, in Bottles
Supplier: VWR Chemicals
Cold Filterable Vegetable Peptone Broth in BPCs (1L bag) 1 * 1 L
Supplier: THERMO FISHER DIAGNOSTICS
PLATE COUNT AGAR 1 * 100 g
Supplier: LIOFILCHEM
Culti-Loops™ <i>Providencia alcalifaciens</i> ATCC™ 51902™
Supplier: THERMO FISHER DIAGNOSTICS
Antibiotic assay discs
Supplier: LIOFILCHEM
Quanti-CultiControl freeze-dried microorganism strains derived from ATCC® cultures
Supplier: LIOFILCHEM
Antibiotic Sensitivity Discs
Supplier: HIMEDIA