Vials
Vials, SN, with write on label, 2,00 ml, Amber, No
Catalog # HEWL5190-4038
Supplier: Agilent
Jump to:
Product Details & Documents
Specifications
- Cap included:No
- Capacity:2,00 ml
- Colour:Amber
- Description:Vials, SN, with write on label
- Seq:06