Orders LinkContactUsLinkComponent
VWR® Separating Funnels, Squibb
1000 ml, 29/32 NS
  532-0028
VWR® Separating Funnels, Squibb
  532-0028
VWR® Separating Funnels, Squibb
VWR® Separating Funnels, Squibb
1000 ml, 29/32 NS
89,30 €
1 * 1 ST
Borosilicate 3.3 glass with NS-PE stopper.
  • With or without graduations
  • Stopcock with PTFE key
  • Very good chemical and temperature resistance
  • Socket size:
    29/32 NS
  • Capacity:
    1000 ml
  • Seq:
    11