VWR® Separating Funnels, Squibb
250 ml, 29/32 NS
Borosilicate 3.3 glass with NS-PE stopper.
- With or without graduations
- Stopcock with PTFE key
- Very good chemical and temperature resistance
- Socket size:29/32 NS
- Capacity:250 ml
- Seq:09