UN CONTAINER 10,9 LITER 1 * 600 Pce
Catalog # HKPLECEM1000191
Supplier: HK PLASTICS
Frequently Bought Together
VWR®, Measuring Beakers with Handle, PMP TPX
Supplier: VWR Collection
VWR®, Universal Filter Tips
Supplier: VWR Collection
Jars, wide mouth, conical, graduated
Supplier: LAMAPLAST
Accessories for single and multi channel pipettes
Supplier: Sartorius
VWR®, Measuring Beakers with Handle
Supplier: VWR Collection
VWR®, Test Tubes Racks
Supplier: VWR Collection
Screw cap for laboratory bottles, GLS 80®, quick release, DURAN®
Supplier: DWK Life Sciences
Filter tips, Thermo Scientific™ ClipTip™
Supplier: Thermo Fisher Scientific
Screw-in tube fittings
Supplier: Bohlender
Storage Bags
Supplier: NEOLAB
Industrial scoops, Plastilab®
Supplier: KARTELL
VWR®, Dispenser Tips, Universal
Supplier: VWR Collection
Container, Securitainer®
Supplier: COVERA
Test tube racks
Supplier: KARTELL
TIP 50µL STERILE FLTRED 1 * 960 items
Supplier: Beckman Coulter
DISPOSABLE FILTER-TIPS 1500 µL, RACKED (8 X 128) 1 * 1 items
Supplier: QIAGEN