ContactUsLinkComponent
23 COMPOSES DANS HNO3 3% ,HF 0,5% Al 1200mg/l Sb 200mg/l As 400mg/l Ag 400mg/l Be 120mg/l Cd 40mg/l Co 1200mg/l Cr 400mg/l Fe 1200mg/l Mn 1200mg/l Hg 40mg/l Ni 400mg/l Pb 400mg/l Cu 1200mg/l Se 400mg/l Sn 40mg/l Tl 8 2 1 * 500 mL
Catalog # CMPL6713.K4.3N05F
23 COMPOSES DANS HNO3 3% ,HF 0,5% Al 1200mg/l Sb 200mg/l As 400mg/l Ag 400mg/l Be 120mg/l Cd 40mg/l Co 1200mg/l Cr 400mg/l Fe 1200mg/l Mn 1200mg/l Hg 40mg/l Ni 400mg/l Pb 400mg/l Cu 1200mg/l Se 400mg/l Sn 40mg/l Tl 8 2 1 * 500 mL
Catalog # CMPL6713.K4.3N05F
Frequently Bought Together