Orders LinkContactUsLinkComponent
Adapters, expansion
Adapters, expansion
Expansion, 34/35 NS, 45/40 NS
Catalog # WITG0.510.019
Supplier:   witeg Labortechnik
Adapters, expansion
Catalog # WITG0.510.019
Supplier:  witeg Labortechnik
Adapters, expansion
Adapters, expansion
Expansion, 34/35 NS, 45/40 NS
Price
52,30 €
Log in for availability
1 * 1 ST
Product Details & Documents
Borosilicate glass 3.3
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Expansion adapters.
Certifications: According to DIN 12257.
Specifications
  • Cone size:
    34/35 NS
  • Socket size:
    45/40 NS
  • Type:
    Expansion
  • Seq:
    03