Tanks, rectangular, LDPE, LaboPlast®
50 L, Seamless containers, rectangular shaped, 1000×320×160 mm
BURK3440-0505
: BURKLE
These seamless rectangular tanks feature optimum ratio of wall thickness to filling load.
- Linear LDPE, transparent
- Rupture-proof
- Capacity:50 L
- Description:Seamless containers, rectangular shaped
- Seq:05
- W×D×H:1000×320×160 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



