ContactUsLinkComponent
Tube capillary microhaematocrit 75?l sodium-heparinized interior 1.1 -1.2 mm, exterior 1.5-1.6mm,made of high grade quality glass in compliance with din iso 12 772,length: approx. 75 mm ± 0.5 mm wall thickness: approx. 0.2 ± 0.025 mm sodium-heparini 1 * 100 items
Catalog # MARI2901000
Supplier:  MARIENFELD
Tube capillary microhaematocrit 75?l sodium-heparinized interior 1.1 -1.2 mm, exterior 1.5-1.6mm,made of high grade quality glass in compliance with din iso 12 772,length: approx. 75 mm ± 0.5 mm wall thickness: approx. 0.2 ± 0.025 mm sodium-heparini 1 * 100 items
Catalog # MARI2901000
Supplier:  MARIENFELD
Frequently Bought Together