ContactUsLinkComponent
Volumetric flasks with one circular mark, 20 ml, +/- 0,04 ml, class A ,borosilicate glass 3.3, clear glass, blue graduated, with ground jointand polystopper, NS 10/19, 10 pcs./carton, MARIENFELD SUPERIOR GTIN: 4250317305701 Customs tariff no.: 701720 1 * 10 items
Catalog # MARI2332201
Supplier:  MARIENFELD
Volumetric flasks with one circular mark, 20 ml, +/- 0,04 ml, class A ,borosilicate glass 3.3, clear glass, blue graduated, with ground jointand polystopper, NS 10/19, 10 pcs./carton, MARIENFELD SUPERIOR GTIN: 4250317305701 Customs tariff no.: 701720 1 * 10 items
Catalog # MARI2332201
Supplier:  MARIENFELD
Frequently Bought Together