ContactUsLinkComponent
Burette 50:0.1ml CL.as with ns glassplug clear glass acc. to mohr,made of borosilicate glass 3.3 in compliance with din en iso 385 v ery good chemical resistance minimal thermal expansion high resistance to thermal shocks 1 * 2 items
Catalog # MARI2422031
Supplier:  MARIENFELD
Burette 50:0.1ml CL.as with ns glassplug clear glass acc. to mohr,made of borosilicate glass 3.3 in compliance with din en iso 385 v ery good chemical resistance minimal thermal expansion high resistance to thermal shocks 1 * 2 items
Catalog # MARI2422031
Supplier:  MARIENFELD
Frequently Bought Together