ContactUsLinkComponent
RECOMBINANT/NATIVE PROTEIN, ANSA (ESCHERICHIA COLI) RECOMBINANT PROTEIN, ESCHERICHIA COLI ANSA (NP_416281, 1 A.A. - 338 A.A.) FULL-L ENGTH RECOMBINANT PROTEIN WITH HIS TAG EXPRESSED IN ESCHERICHIA COLI. 1 * 100 µG
Catalog # ABNOP5008
Supplier:  Abnova
RECOMBINANT/NATIVE PROTEIN, ANSA (ESCHERICHIA COLI) RECOMBINANT PROTEIN, ESCHERICHIA COLI ANSA (NP_416281, 1 A.A. - 338 A.A.) FULL-L ENGTH RECOMBINANT PROTEIN WITH HIS TAG EXPRESSED IN ESCHERICHIA COLI. 1 * 100 µG
Catalog # ABNOP5008
Supplier:  Abnova