Orders LinkContactUsLinkComponent
Gel drying Frames™
Gel drying Frames™
Gel drying frame, 140×140 mm
Catalog # DIVEAB-110760
Supplier:   DIVERSIFIED BIOTECH
Gel drying Frames™
Catalog # DIVEAB-110760
Gel drying Frames™
Gel drying Frames™
Gel drying frame, 140×140 mm
Price
363,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents
Gel drying frames allow researchers for easy handling and crack free dry gels without the use of heat/vacuum dryers.
Delivery information: The complete kit includes 2 open frames, 2 solid back plates, 12 side clips and 50 sheets of pre-cut cellophane.
Specifications
  • Description:
    Gel drying frame, 140×140 mm
  • Seq:
    01