ContactUsLinkComponent
KIT, ENA SCREEN ELISA KIT, ENA SCREEN ELISA IS AN INDIRECT SOLID PHASE ELISA FOR THE QUALITATIVE SCREENING OF IGG CLASS AUTOANTIBODI ES AGAINST EXTRACTABLE NUCLEAR ANTIGENS (ENA) IN HUMAN SERUM OR PLASMA. 1 * 1 KIT
Catalog # ABNOKA1106
Supplier:  Abnova
KIT, ENA SCREEN ELISA KIT, ENA SCREEN ELISA IS AN INDIRECT SOLID PHASE ELISA FOR THE QUALITATIVE SCREENING OF IGG CLASS AUTOANTIBODI ES AGAINST EXTRACTABLE NUCLEAR ANTIGENS (ENA) IN HUMAN SERUM OR PLASMA. 1 * 1 KIT
Catalog # ABNOKA1106
Supplier:  Abnova