ContactUsLinkComponent
KIT, ENA-6 PROFILE ELISA KIT, ENA-6-PROFILE ELISA KIT IS AN INDIRECT SOLID PHASE ELISA FOR SEMI-QUANTITATIVE DETERMINATION OF IGG CL ASS AUTOANTIBODIES DIRECTED AGAINST THE EXTRACTABLE NUCLEAR ANTIGENS SS-A (RO), SS-B (LA), SM RNP/SM, SCL-70 AND JO-1 1 * 1 KIT
Catalog # ABNOKA1105
Supplier:  Abnova
KIT, ENA-6 PROFILE ELISA KIT, ENA-6-PROFILE ELISA KIT IS AN INDIRECT SOLID PHASE ELISA FOR SEMI-QUANTITATIVE DETERMINATION OF IGG CL ASS AUTOANTIBODIES DIRECTED AGAINST THE EXTRACTABLE NUCLEAR ANTIGENS SS-A (RO), SS-B (LA), SM RNP/SM, SCL-70 AND JO-1 1 * 1 KIT
Catalog # ABNOKA1105
Supplier:  Abnova