Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Accessories

E 362 Blind stopper for the sealing of fittings to load carriers, W×D×H: 222×471×185 mm
Catalog # 141-0839
Supplier:   MIELE
Accessories
Catalog # 141-0839
Supplier:  MIELE
Accessories
Accessories

E 362 Blind stopper for the sealing of fittings to load carriers, W×D×H: 222×471×185 mm

Price
10,45 €
Log in for availability
1 * 1 ST
Product Details & Documents
  • For lower and upper baskets
  • With 7 x 7 mm perforations
  • Stainless steel
Specifications
  • Description:
    E 362 Blind stopper for the sealing of fittings to load carriers, W×D×H: 222×471×185 mm
  • Seq:
    13