Order Entry
Belgium
Orders LinkContactUsLinkComponent
pH buffer solutions

pH buffer solutions

pH buffer standard solution with certificate of analysis, 125 ml, 9,21, 1
Catalog # HACHLZW9462.99
Supplier:   Hach
pH buffer solutions
Catalog # HACHLZW9462.99
Supplier:  Hach
CAS Number:  
pH buffer solutions
pH buffer solutions

pH buffer standard solution with certificate of analysis, 125 ml, 9,21, 1

CAS Number:  
Price
16,10 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications

Specifications

  • Pk:
    1
  • Description:
    pH buffer standard solution with certificate of analysis, 125 ml
  • pH value:
    9,21

Specifications