Orders LinkContactUsLinkComponent
Modules for inoLab® Multi IDS
Modules for inoLab® Multi IDS
ADA 94pH/IDS BNC, pH/mV module for inoLab® Multi 9420/30 IDS for pH/ORP electrodes with BNC- and 4 mm banana plug, incl. mounting accessories
Catalog # 665-0372
Supplier:   WTW
Modules for inoLab® Multi IDS
Catalog # 665-0372
Supplier:  WTW
Modules for inoLab® Multi IDS
Modules for inoLab® Multi IDS
ADA 94pH/IDS BNC, pH/mV module for inoLab® Multi 9420/30 IDS for pH/ORP electrodes with BNC- and 4 mm banana plug, incl. mounting accessories
Price
405,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Integratable adapters for connecting Multi IDS meters with conventional pH/ORP/ISE electrodes.
Documents
Specifications
  • Description:
    ADA 94pH/IDS BNC, pH/mV module for inoLab® Multi 9420/30 IDS for pH/ORP electrodes with BNC- and 4 mm banana plug, incl. mounting accessories
  • Seq:
    02