Storage Sacks
Approx. 78 bags/pack, 0,06 mm, 800×1400 mm
Catalog # LAMA129
Supplier: LAMAPLAST
Jump to:
Product Details & Documents
LDPE, neutral storage bags.
- With bottom weld
- Can be used for foodstuffs
Specifications
- Description:Approx. 78 bags/pack
- Dimensions:800×1400 mm
- Seq:10
- Thickness:0,06 mm



