Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Storage Sacks

Approx. 109 bags/pack, 0,06 mm, 750×1100 mm
  LAMA128
 :   LAMAPLAST
Storage Sacks
  LAMA128
 :  LAMAPLAST
Storage Sacks
Storage Sacks

Approx. 109 bags/pack, 0,06 mm, 750×1100 mm

 
244,00 €
   
1 * 10 kg
 

LDPE, neutral storage bags.

  • With bottom weld
  • Can be used for foodstuffs
 
  • Description:
    Approx. 109 bags/pack
  • Dimensions:
    750×1100 mm
  • Seq:
    09
  • Thickness:
    0,06 mm