Order Entry
Belgium
Orders LinkContactUsLinkComponent
Conductivity standards

Conductivity standards

1382 ppm TDS calibration standard, 500 mL
Catalog # JENW16156-81
Supplier:   Cole-Parmer
Conductivity standards
Catalog # JENW16156-81
Supplier:  Cole-Parmer
CAS Number:  
Conductivity standards

1382 ppm TDS calibration standard, 500 mL

CAS Number:  
Price
101,00 €
Log in for availability
1 * 500 mL
Product Details & Documents

Standard solutions are used for calibrating conductivity cells.

Specifications

Specifications

  • Pk:
    500 mL
  • Description:
    1382 ppm TDS calibration standard

Specifications