Order Entry
Belgium
Orders LinkContactUsLinkComponent
Redox standards

Redox standards

200 mV redox standard, 500 mL
Catalog # JENW90238-49
Supplier:   Cole-Parmer
Redox standards
Catalog # JENW90238-49
Supplier:  Cole-Parmer
CAS Number:  
Redox standards

200 mV redox standard, 500 mL

CAS Number:  
Price
251,00 €
Log in for availability
1 * 500 mL
Product Details & Documents
Specifications

Specifications

  • Pk:
    500 mL
  • Description:
    200 mV redox standard

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.